Anti-IL27RA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-2711R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Interleukin 27 Receptor Alpha
- Antigen symbol:IL27RA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:9466
- Antigen synonyms:WSX1|Interleukin-27 receptor subunit alpha|zcytor1|IL27R|UNQ296/PRO336|CRL1|TCCR|IL-27RA|Cytokine receptor WSX-1|IL-27 receptor subunit alpha
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:35-80/636
- Purification:Purified by Protein A
- Pk:100 µl
Receptor for IL27. Requires IL6ST/gp130 to mediate signal transduction in response to IL27. This signaling system acts through STAT3 and STAT1. Involved in the regulation of Th1-type immune responses. Also appears to be involved in innate defense mechanisms.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL27RA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat