Anti-PSENEN Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-0771R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Presenilin Enhancer Gamma Secretase Subunit
- Antigen symbol:PSENEN
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:55851
- Antigen synonyms:PEN2|MDS033|Presenilin enhancer protein 2|Gamma-secretase subunit PEN-2|PEN-2|MSTP064
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:129-176/478
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Essential subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP (beta-amyloid precursor protein). Probably represents the last step of maturation of gamma-secretase, facilitating endoproteolysis of presenilin and conferring gamma-secretase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PSENEN
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat