Anti-CEBPA Rabbit Polyclonal Antibody (Cy5.5®)
BOSSBS-3061R-CY5.5
: Bioss
- Antibody type:Primary
- Antigen name:CCAAT/enhancer binding protein (C/EBP), alpha
- Antigen symbol:CEBPA
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1050
- Antigen synonyms:C/EBP alpha|CCAAT/enhancer-binding protein alpha|CEBPA
- Modification:phospho-Thr226
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:200-250/358
- Purification:Purified by Protein A
- Pk:100 µl
C/EBP is a DNA-binding protein that recognizes two different motifs: the CCAAT homology common to many promoters and the enhanced core homology common to many enhancers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CEBPA
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: