Order Entry
United Kingdom
ContactUsLinkComponent
Anti-COX7A2 Rabbit Polyclonal Antibody (Cy3®)
Anti-COX7A2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-2562R-CY3
Supplier:  Bioss
Anti-COX7A2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-2562R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome c oxidase subunit VIIa polypeptide 2 (liver)
  • Antigen symbol:
    COX7A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1347
  • Antigen synonyms:
    COXVIIAL|COX7AL|COX7AL1|COXVIIa-L|VIIAL
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Cytochrome c oxidase, the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of three catalytic subunits encoded by mitochondrial genes, and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, while the nuclear-encoded subunits may function in the regulation and assembly of the complex. This nuclear gene encodes polypeptide 2 (liver isoform) of subunit VIIa, with this polypeptide being present in both muscle and non-muscle tissues. In addition to polypeptide 2, subunit VIIa includes polypeptide 1 (muscle isoform), which is present only in muscle tissues, and a related protein, which is present in all tissues. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 4 and 14. [provided by RefSeq, Oct 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX7A2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: