Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CCR9 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-1788R-CY5
 :  Bioss
Anti-CCR9 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-1788R-CY5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Chemokine (C-C motif) receptor 9
  • Antigen symbol:
    CCR9
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10803
  • Antigen synonyms:
    CDw199|GPR-9-6|CC-CKR-9|GPR28
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    107-127/369
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Receptor for chemokine SCYA25/TECK. Subsequently transduces a signal by increasing the intracellular calcium ions level. Alternative coreceptor with CD4 for HIV-1 infection.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCR9
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat