Anti-GGT7 Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-13351R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:Gamma-Glutamyltransferase 7
- Antigen symbol:GGT7
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:2686
- Antigen synonyms:Gamma glutamyltransferase 4|Gamma glutamyltransferase like 3|GGTL3|D20S101|Gamma-glutamyltransferase 7|GGT4|Gamma glutamyltranspeptidase 7|GGT7_HUMAN.|Gamma glutamyltransferase 7|dJ18C9.2|GGT 7|GGTL5|Gamma glutamyltransferase like 5|Gamma glutamyltranspeptidase like 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene is a member of a gene family that encodes enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. The protein encoded by this gene consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis. [provided by RefSeq, Jul 2008].
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: GGT7 heavy chain
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat