- Antibody type:Primary
- Antigen name:Epidermal growth factor-like protein 6
- Antigen symbol:EGFL6
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:25975
- Antigen synonyms:MAEG|W80
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene encodes a member of the epidermal growth factor(EGF) repeat superfamily. Members of this superfamily arecharacterized by the presence of EGF-like repeats and are ofteninvolved in the regulation of cell cycle, proliferation, anddevelopmental processes. The gene product contains a signalpeptide, suggesting that it is secreted; an EGF repeat regionconsisting of 4 complete EGF-like repeats and 1 partial EGF-likerepeat, 3 of which have a calcium-binding consensus sequence; anarg-gly-asp integrin association motif; and a MAM domain, which isbelieved to have an adhesive function. This gene is expressed earlyduring development, and its expression has been detected in lungand meningioma tumors. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EGFL6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat