Anti-ACAA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-12566R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-CoA Acyltransferase 1
- Antigen symbol:ACAA1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:30
- Antigen synonyms:THIO|ACAA|PTHIO
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ACAA1 is a 424 amino acid member of the thiolase family of enzymes and is involved in lipid metabolism. Localized to the peroxisome, ACAA1 catalyzes the conversion of acyl-CoA and acetyl-CoA to 3-oxoacyl-CoA in the fatty acid oxidation pathway. ACAA1 shows high enzymatic activity in liver, kidney, intestine and white adipose tissue in rats, where it exists as two types, namely type A and type B. Human ACAA1 shares 86% amino acid identity with its rat counterpart, suggesting a conserved function for ACAA1 among different species.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACAA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat