Anti-CDC16 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-12486R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Cell Division Cycle 16
- Antigen symbol:CDC16
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8881
- Antigen synonyms:Anaphase-promoting complex subunit 6|ANAPC6|CUT9|APC6|Cell Division Cycle protein 16 homolog
- Modification:phospho-Ser560
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of 'Lys-11'-linked polyubiquitin chains and, to a lower extent, the formation of 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDC16
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: