Specifications
- Antibody type:Primary
- Antigen name:NEDD8 Activating Enzyme E1 Subunit 1
- Antigen symbol:NAE1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8883
- Antigen synonyms:APPBP1|A-116A10.1|ula-1|HPP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
APPBP1 is a member of the ubiquitin-activating E1 family. In fetal tissues APPBP1 is widely expressed and in adult tissues it is expressed throughout the brain. APPBP1 is a cell membrane associated protein and functions as the regulatory subunit in a heterodimer with UBA3. The APPBP1/UBA3 complex binds to and activates NEDD8, a ubiquitin-like protein involved in signal transduction, cell proliferation and development. This suggests that APPBP1 affects a variety of cellular functions. In addition, APPBP1 is essential for cell cycle progression through the S/M checkpoint. More specifically, it inhibits the entry into S phase and promotes entry into M phase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NAE1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat