Specifications
- Antibody type:Primary
- Antigen name:5-Hydroxytryptamine receptor 7
- Antigen symbol:5HT7R
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:5-HT7|HTR7|SR-7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Serotonin (also designated 5-hydroxytryptamine or 5-HT) is a molecule that functions as a neurotransmitter, a hormone and a mitogen, and it is predominantly expressed in the gut, platelets and central nervous system (CNS). In the CNS, serotonin modulates several processes, including anxiety, sleep, appetite, behavior and drug abuse. In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. SR-3 is a ligand-gated ion channel, whereas all other known serotonin receptor subtypes are G protein-coupled receptors. SR-4 mediates widespread effects in central and peripheral nervous systems. SR-7 belongs to the superfamily of G protein-coupled receptors. The gene which encodes SR-7 maps to human chromosome 10q21-q24.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: 5HT7R
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat