- Antibody type:Primary
- Antigen name:HCLS1 binding protein 3
- Antigen symbol:HS1BP3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HS1-BP3|ETM2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
HS1BP3 is a 392 amino acid protein that contains one PX (phox homology) domain, a leucine zipper, immunoreceptor tyrosine-based inhibitory motif-like motifs and multiple proline-rich regions. Expressed primarily in brain, HS1BP3 is encoded by a gene mapping to human chromosome 2p24.1. The gene encoding HS1BP3 is frequently mutated in familial essential tremor, a disorder characterized by kinetic tremor the the hands, voice or head, though there is no correlation to Parkinson disease. HS1BP3 interacts with HAX-1’s SH3 domain, and may also play a role in the regulation of catecholamine and serotonin metabolism. Acting as a regulator of IL-2 signaling, HS1BP3 is likely involved in lymphocyte activation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HS1BP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat