Anti-PTPZ Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BOSSBS-11327R-PE
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase zeta
- Antigen symbol:PTPZ
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:5803
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:460-500/2315
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades (By similarity).
Recommended Dilutions: Flow Cytometry: 1:20-100
Type: Primary
Antigen: PTP zeta
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat