Anti-SYT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1136R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Synaptotagmin 1
- Antigen symbol:SYT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:127833
- Antigen synonyms:Syt1|Synaptotagmin-2|Synaptotagmin II|p65|SYT2|FLJ42519|SytI|Synaptotagmin-1|synaptotagmin 2|Synaptotagmin I|SytII
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).[supplied by OMIM]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SYT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat