- Antibody type:Primary
- Antigen name:Pleiomorphic Adenoma Gene-Like 1 Protein
- Antigen symbol:PLAGL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PLAL1_HUMAN|DKFZp781P1017|Lost on transformation 1|Pleiomorphic adenoma-like protein 1|Pleiomorphic adenoma gene like 1|LOT1|ZAC1|ZAC tumor supressor|Pleiomorphic adenoma gene like protein 1|Tumor supressor ZAC|LOT-1|MGC126275|PLAG like 1|Zinc finger protein PLAGL1.|Lost on Transformation|MGC126276|ZAC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene encodes a C2H2 zinc finger protein with transactivation and DNA-binding activities. It has been shown to have anti-proliferative properties, and thus thought to function as a tumor suppressor. In addition, overexpression of this gene during fetal development is believed to underlie the rare disorder, transient neonatal diabetes mellitus (TNDM). This gene is imprinted, with preferential expression of the paternal allele in many tissues, however, biallelic expression has been noted in peripheral blood leucocytes. A recent study reports that tissue-specific imprinting results from variable utilization of monoallelic and biallelic promoters. Many transcript variants differing in the 5' UTR and encoding two different isoforms, have been found for this gene. [provided by RefSeq, Oct 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLAGL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat