Specifications
- Antibody type:Primary
- Antigen name:Cadherin 18
- Antigen symbol:CDH18
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Cadherin 18|Cadherin 14|Cadherin18|CDH18|CDH14|Cadherin-14|Cadherin14
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding events that are critical to the maintenance of cell structure and morphogenesis. EY-cadherin, also known as CDH18 (cadherin 18), CDH14 (cadherin 14), CDH24 or CDH14L, is a 790 amino acid single-pass type I membrane protein that contains five cadherin domains. One of several members of the cadherin superfamily, EY-cadherin functions as a type II classical cadherin that is expressed specifically in the central nervous system (CNS), where it plays a role in cell-cell binding events. Specifically, EY-cadherin is thought to be involved in axon guidance and outgrowth, as well as synaptic adhesion within the CNS. EY-cadherin contains a highly conserved C-terminal domain characteristic of all cadherins, but lacks the HAV cell adhesion sequence that is specific to type I cadherins. The gene encoding EY-cadherin is located within a region on chromosome five that is commonly deleted in carcinomas, implicating EY-cadherin as a potential tumor suppressor.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDH18
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: