Anti-KIF15 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-10693R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Kinesin Family member 15
- Antigen symbol:KIF15
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:56992
- Antigen synonyms:KNSL7|HKLP2|NY-BR-62
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
KIF15 belongs to the kinesin-like protein family and contains 1 kinesin-motor domain. KIF15 is the plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KIF15
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat