Anti-PROC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-10486R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Protein C light chain
- Antigen symbol:PROC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Vitamin K-dependent protein C light chain.|proC|APC|Vitamin K-dependent protein C|Protein C|Protein C inactivator of coagulation factors Va and VIIIa|PROC|PROC_HUMAN|Vitamin K dependent protein C|EC 3.4.21.69|PC|Autoprothrombin IIA|Vitamin K dependent protein C precursor|Anticoagulant protein C|PROC1|Blood coagulation factor XIV
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a vitamin K-dependent plasma glycoprotein. The encoded protein is cleaved to its activated form by the thrombin-thrombomodulin complex. This activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Mutations in this gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.[provided by RefSeq, Dec 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: protein C light chain
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat