Anti-ATP-Citrate Lyase Mouse Monoclonal Antibody
BWRLMB0070
- Antibody type:Primary
- Antigen symbol:ATP-Citrate Lyase
- Clonality:Monoclonal
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids C-terminus of Human ATP-Citrate Lyase.
This antibody detects endogenous levels of ATP-Citrate Lyase (C-terminus) and does not cross-react with related proteins
Type: Primary
Antigen: ATP-Citrate Lyase
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human