Order Entry
United Kingdom
ContactUsLinkComponent
Anti-p-MEF-2 Rabbit Polyclonal Antibody [clone: S408]
Anti-p-MEF-2 Rabbit Polyclonal Antibody [clone: S408]
  BWRLBS4119
Anti-p-MEF-2 Rabbit Polyclonal Antibody [clone: S408]
  BWRLBS4119

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p-MEF-2
  • Clonality:
    Polyclonal
  • Clone:
    S408
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic phosphopeptide derived from human MEF-2 around the phosphorylation site of Serine 408.

p-MEF-2 (S408) pAb detects endogenous levels of MEF-2 protein only when phosphorylated at Ser408.

Type: Primary
Antigen: p-MEF-2
Clonality: Polyclonal
Clone: S408
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat