Anti-PKA Ialpha reg Rabbit Polyclonal Antibody [clone: Q304 ]
Catalog # BWRLBS1926
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKA I alpha reg
- Clonality:Polyclonal
- Clone:Q304
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 271-320 of Human PKA Ialpha reg.
PKA I? reg (Q304) pAb detects endogenous levels of PKA I? reg protein
Type: Primary
Antigen: PKA Ialpha reg
Clonality: Polyclonal
Clone: Q304
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat