Anti-CB2 Rabbit Polyclonal Antibody [clone: R334]
BWRLBS2615
- Antibody type:Primary
- Antigen symbol:CB2
- Clonality:Polyclonal
- Clone:R334
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids C-terminus of Human CB2.
CB2 (R334) pAb detects endogenous levels of CB2 protein.
Type: Primary
Antigen: CB2
Clonality: Polyclonal
Clone: R334
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat