Anti-Cadherin-8 Rabbit Polyclonal Antibody [clone: A519]
Catalog # BWRLBS2578
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cadherin-8
- Clonality:Polyclonal
- Clone:A519
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 500-550 of Human Cadherin-8.
Cadherin-8 (A519) pAb detects endogenous levels of Cadherin-8 protein.
Type: Primary
Antigen: Cadherin-8
Clonality: Polyclonal
Clone: A519
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat