Anti-ZIP-kinase Rabbit Polyclonal Antibody [clone: K261]
Catalog # BWRLBS3724
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ZIP-kinase
- Clonality:Polyclonal
- Clone:K261
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 231-280 of Human ZIP-kinase.
ZIP-kinase (K261) pAb detects endogenous levels of ZIP-kinase protein.
Type: Primary
Antigen: ZIP-kinase
Clonality: Polyclonal
Clone: K261
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat