Anti-GCS-beta-1 Rabbit Polyclonal Antibody [clone: V21]
Catalog # BWRLBS1158
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GCS- beta-1
- Clonality:Polyclonal
- Clone:V21
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human GCS-beta-1.
GCS-beta-1 (V21) pAb detects endogenous levels of Guanylate Cyclase ?1/3 protein.
Type: Primary
Antigen: GCS-beta-1
Clonality: Polyclonal
Clone: V21
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat