Chlorine water ∼0,4% Cl saturated solution USP Chlorine Test Solution
Catalog # SPCMC-205-500ML
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
:
UN:1760
:GHS05
:GHS07
CAS Number:7782-50-5
ADR:8 III