Order Entry
United Kingdom
ContactUsLinkComponent
Dialysis tubing, Visking®
  MEDCDTV.12000.10
 :  MEDICELL
Dialysis tubing, Visking®
  MEDCDTV.12000.10
 :  MEDICELL
  • Ø:
    31,7 mm
  • MWCO:
    12 - 14 kD
  • Vol./ length:
    — ml/cm
  • Environmentally Preferable:
  • Pk:
    15 m
Regenerated cellulose (RC).
  • Contain small amounts of glycerol
  • Dry packaged
  • Stable at pH range 5 to 9
  • Autoclavable up to +120 °C (max. 15 min, completely submersed in dist. water)
  • No non-specific protein binding (<1 ng/g)
For laboratory analysis and ultra filtration.
Storage stability in original packaging 2 years.