Sodium dodecyl sulphate (SDS) 20% in aqueous solution , 99.5%
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Density:1.03 g/mL (20 °C)
Molecular Weight:288.38 g/mol
Merck Index:14,08636
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:>99.5%
Specification Test Results
Frequently Bought Together