Sodium mercaptoacetate (sodium thioglycolate)
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt
CAS Number:367-51-1
:
Formula:HSCH₂COONa
MDL Number:MFCD00043386
Molecular Weight:114.1 g/mol
UN:3335
ADR:9 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together