Order Entry
United Kingdom
ContactUsLinkComponent
Alkaline Batteries
  DIALEA6752
undefined
Alkaline Batteries
  DIALEA6752
  • Type:
    Mignon AA
  • Description:
    Alkaline battery
  • Environmentally Preferable:
  • Pk:
    10
Duracell Industrial batteries are designed for industrial-strength power. These AA batteries are ideal for use in torches, safety equipment, communications devices, telemeters, medical instruments and more. They are designed for extreme usage.
  • Capable of operation in temperatures from –20 to 54 °C
  • Storage up to seven years