Membrane filters, cellulose acetate, Whatman™
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
- Pure cellulose acetate composition, minimising sample contamination
- Hydrophilic, for use with aqueous and alcoholic media
- High solvent resistance, particularly to low molecular weight alcohols
- High physical strength and a heat resistance up to 180 °C
- Can be sterilized by most common methods without sacrificing membrane integrity
Due to their high chemical resistance, cellulose acetate membrane filters reliably prepare liquid scintillation cocktail for scintillation counter measurement.
High chemical resistance membranes.
Additionally, the highly heat resistant Whatman cellulose acetate membranes can filter hot liquids and hot gases.
- Catalog No:
- 516-5016
- 516-5019
- 514-2362
- 514-0103
- 514-2353
- 514-2363
- 514-2322
- 516-5020
- 514-2350
- 516-5015
- 516-5018
- 514-2365
- 514-2351
- 514-2354
- 514-2352
- 514-0100
- 514-0769
- 514-0770
- 514-2361
- Type:
- OE 67
- OE 66
- OE 66
- WCA
- OE 67
- OE 66
- ST 69
- OE 66
- OE 67
- OE 67
- OE 66
- OE 66
- OE 67
- OE 67
- OE 67
- ST 68
- WCA
- WCA
- OE 66
- Pore size:
- 0,45 µm
- 0,20 µm
- 0,20 µm
- 0,20 µm
- 0,45 µm
- 0,20 µm
- 1,20 µm
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,80 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- Thickness:
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 140 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 115 µm
- 140 µm
- 115 µm
- 115 µm
- 115 µm
- Water flow rate:
- 12 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 16 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 12 s/100 ml/12,5 cm²
- 26 s/100 ml/12,5 cm²
- Airflow rate:
- 25 ml/min/cm² (3 mbar)
- 25 ml/min/cm² (3 mbar)
- 25 ml/min/cm² (3 mbar)
- 25 ml/min/cm² (3 mbar)
- 25 ml/min/cm² (3 mbar)
- Max. temperature:
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
Frequently Bought Together