Inlet liners, SPME-GC, Supelco®
Supplier: Merck
Certificates
About this item
SPME-GC inlet liners are designed for optimum sample introduction for specific injection techniques that are suitable for Agilent, Perkinelmer, Shimadzu, Finnigan, Varian and Thermo scientific equipped with split/splitless inlets.
- Deactivated (silanized) glass inlet liner for capillary injection ports
Frequently Bought Together