Gas traps and connector kits, Click-On Inline Super Clean®
REST22458
: Restek
- Description:Gas trap and connector kit, includes two 1/4" SS connectors and one oxygen/moisture/hydrocarbon triple trap
- Seq:02
- Environmentally Preferable:
- Pk:1 KIT
This gas purification system with triple gas trap is ideal for purifying carrier gas and it contains oxygen, moisture, and hydrocarbon scrubbers in one cartridge.
- Click-On fittings for easy, leak-tight cartridge changes
- High-purity output ensures 99.9999% pure gas
Click-On adaptor connectors allow cartridges to be exchanged without introducing oxygen, moisture and hydrocarbons. Spring-loaded check valves seal when a filter is removed and open only when a new filter has been locked in place.