Order Entry
United Kingdom
ContactUsLinkComponent
FID jet removal tool
FID jet removal tool
Catalog # REST22328
Supplier:  Restek
undefined
FID jet removal tool
Catalog # REST22328
Supplier:  Restek
Specifications
  • Description:
    FID jet removal kit
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
These FID jet removal tool for Agilent 5890/6850/6890/7890 FID.
  • Securely grips jet in socket for easy removal or installation
  • Unique, ergonomic handle and easy to hold