Order Entry
United Kingdom
ContactUsLinkComponent
Ion exchange resins, Diaion® HP20
  46488.A1
 :  
Ion exchange resins, Diaion® HP20
  46488.A1
 :  
  • Particle Size:
    0,25 mm
  • Pk:
    1 kg
  • Description:
    Synthetic adsorbent ion exchange resin, Diaion® HP20, highly porous type
  • Environmentally Preferable:
Diaion® HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.