Order Entry
United Kingdom
ContactUsLinkComponent
Cylinder holder, wall mounted
Cylinder holder, wall mounted
Catalog # REST21333
Supplier:  Restek
undefined
Cylinder holder, wall mounted
Catalog # REST21333
Supplier:  Restek
Specifications
  • Type:
    Cylinder holder, single
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This holder is designed to prevent free-standing gas cylinders from tipping over and injuring personnel.
  • Cast aluminum holder can be secured to a wall or the side of a work bench
  • Each mount will secure a cylinder 101,6 to 381 mm in diameter