Polycarbonate erlenmeyer flasks with screw caps, Corning®
Supplier: Corning
Certificates
About this item
PC, optically clear.
- Baffled or plain bottom options in all sizes (125 ml to 3 L)
- Moulded-in graduations for accuracy
- Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
- Individually packaged and radiation sterilised for ease of use
- Certified non-pyrogenic
Corning® baffled and plain Erlenmeyer flasks are ideal for shaker culture applications and storage.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
- Catalog No:
- 734-1903
- 734-1889
- 734-1902
- 734-4204
- 734-1888
- 734-4203
- 734-4202
- 734-4201
- 734-4200
- 734-4199
- 734-1820
- 734-4197
- 734-2759
- 734-2758
- 734-4198
- 734-1887
- 734-1833
- 734-1886
- 734-1905
- 734-1832
- 734-1885
- 734-1904
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
Specifications
Related Information
Aseptic Transfer Caps
Product Information
Frequently Bought Together