Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CD84 Mouse Monoclonal Antibody (CF488A) [clone: 153-4D9]
Anti-CD84 Mouse Monoclonal Antibody (CF488A) [clone: 153-4D9]
Supplier:  Biotium
Certificates
Anti-CD84 Mouse Monoclonal Antibody (CF488A) [clone: 153-4D9]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881049-100
  • BNC881049-500
  • Antigen symbol:
  • CD84
  • CD84
  • Antigen synonyms:
  • LY9B|Signaling lymphocytic activation molecule 5 (SLAMF5)|hCD84|Cell surface antigen MAX.3|SLAM family member 5|Hly9-beta|Leucocyte differentiation antigen CD84
  • LY9B|Signaling lymphocytic activation molecule 5 (SLAMF5)|hCD84|Cell surface antigen MAX.3|SLAM family member 5|Hly9-beta|Leucocyte differentiation antigen CD84
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone 153-4D9
  • Clone 153-4D9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 398093
  • 398093
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together