Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CALP CNN1/832]
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CALP CNN1/832]
Supplier:  Biotium
Certificates
Anti-CNN1 Mouse Monoclonal Antibody (CF594) [clone: CALP CNN1/832]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940833-500
  • BNC940833-100
  • Antigen symbol:
  • CNN1
  • CNN1
  • Antigen name:
  • Calponin-1
  • Calponin-1
  • Antigen synonyms:
  • Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
  • Calponin-1|Calponin 1 basic smooth muscle|Sm Calp|CNN1|SMCC|Cnn1|Calponin H1 smooth muscle
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone CALP CNN1/832
  • Clone CALP CNN1/832
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 465929
  • 465929
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together