Anti-SLCO2B1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3913R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Organic Anion Transporter Family Member 2B1
- Antigen symbol:Solute carrier organic anion transporter family, member 2B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:11309
- Antigen synonyms:OATPB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:584-597/709
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SLCO2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat