Anti-CKLF Rabbit Polyclonal Antibody
BOSSBS-2394R
: Bioss
- Antibody type:Primary
- Antigen name:Chemokine-like Factor
- Antigen symbol:CKLF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:51192
- Antigen synonyms:CKLF4|C32|CKLF1|HSPC224|CKLF3|CKLF2|UCK-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:30-65/152
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Type: Primary
Antigen: CKLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat