Anti-NKB Rabbit Polyclonal Antibody
BOSSBS-0070R
: Bioss
- Antibody type:Primary
- Antigen name:Neurokinin B
- Antigen symbol:NKB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:6866
- Antigen synonyms:NKB|PRO1155|ZNEUROK1|NKNB
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at –20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Immunogen:81-94/121
- Purification:Purified by Protein A
- Size:100 μl
- Pk:100 µl
Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles (by similarity). Is a critical central regulator of gonadal function.
Recommended dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: NKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat