Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-PODXL Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 2A4]
Catalog # BNUM0483-50
Supplier:  Biotium
Anti-PODXL Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 2A4]
Catalog # BNUM0483-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Podocalyxin
  • Antigen symbol:
    PODX
  • Clonality:
    Monoclonal
  • Clone:
    Clone 2A4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Rabbit,
    Human,
    Rat
  • Environmentally Preferable:
  • Gene ID:
    732423
  • Antigen synonyms:
    PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 2A4
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Rabbit, Rat