About this item
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4 /CD8-. Anaplastic large cell lymphoma is usually CD4 and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: C8/468
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNUB0468-100
- BNUB0468-500
- Antigen symbol:
- CD8A
- CD8A
- Antigen name:
- CD8
- CD8
- Antigen synonyms:
- T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
- T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone C8/468
- Clone C8/468
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 85258
- 85258
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide