Anti-PCNA Mouse Monoclonal Antibody (Purified) [clone: PC10]
Supplier: Biotium
Certificates
About this item
Recognizes a non-histone protein of 36 kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This MAb is excellent for multiple applications.
Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: PC10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chicken, Drosophila, Human, Monkey, Mouse, Pig, Rat, S. cerevisiae), S. pombe, Zebrafish
Specifications
- Catalog No:
- BNUB0467-500
- BNUB0467-100
- Antigen symbol:
- PCNA
- PCNA
- Antigen name:
- Proliferating cell nuclear antigen
- Proliferating cell nuclear antigen
- Antigen synonyms:
- DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|Polymerase delta accessory protein|Cyclin|PCNAR
- DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|Polymerase delta accessory protein|Cyclin|PCNAR
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone PC10
- Clone PC10
- Reactivity:
- Zebrafish, S. cerevisiae, Drosophila, Pig, Monkey, S. pombe, Chicken, Human, Rat, Mouse
- Zebrafish, S. cerevisiae, Drosophila, Pig, Monkey, S. pombe, Chicken, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 147433 , 728886
- 147433 , 728886
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
Specifications
Frequently Bought Together