- Antibody type:Primary
- Antigen name:Beta-2 Microglobulin
- Antigen symbol:B2M
- Clonality:Monoclonal
- Clone:Clone BBM.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Primates,Human
- Environmentally Preferable:
- Gene ID:534255
- Antigen synonyms:Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Recognizes a protein of 12 kDa, identified as microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of an α heavy chain that contains three subdomains (α1, α2, α3) and a non-covalent associating light chain, known as β-2-Microglobulin. β-2-Microglobulin associates with the α3 subdomain of the α heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. The α1 and α2 domains of the α heavy chain form the peptide antigen-binding cleft. Mutations in the β-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes.
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: BBM.1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Non-human primates