Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BAX Mouse Monoclonal Antibody (Biotin) [clone: 2D2]
Anti-BAX Mouse Monoclonal Antibody (Biotin) [clone: 2D2]
Supplier:  Biotium
Certificates
Anti-BAX Mouse Monoclonal Antibody (Biotin) [clone: 2D2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0004-100
  • BNCB0004-500
  • Antigen symbol:
  • BAX
  • BAX
  • Antigen name:
  • Bax
  • Bax
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone 2D2
  • Clone 2D2
  • Reactivity:
  • Monkey, Human
  • Monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 624291
  • 624291
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together