Order Entry
United Kingdom
ContactUsLinkComponent
DNA extraction kits, MPure™ Bacterial DNA extraction kit, MP Biomedicals
DNA extraction kits, MPure™ Bacterial DNA extraction kit, MP Biomedicals
Catalog # ICNA117022600
Supplier:  MP Biomedicals
CAS Number:  
undefined
DNA extraction kits, MPure™ Bacterial DNA extraction kit, MP Biomedicals
Catalog # ICNA117022600
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Application:
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    MPure™ Bacterial DNA Extraction Kit, 48 tests
  • Environmentally Preferable:
  • Description:
    MPure™ Bacterial DNA Extraction Kit, 48 tests

Specifications

About this item

MPure™ Bacterial DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction and purification of genomic DNA from both gram-positive and gram-negative bacteria. Nucleic acids extracted and purified from MPure™ Bacterial DNA Extraction Kit can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.