Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]
Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]
Supplier:  ENZO LIFE SCIENCES
New Product
Certificates
Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]
Supplier:  ENZO LIFE SCIENCES
New Product

Specifications

  • Catalog No:
  • ENZOADISPA470F
  • ENZOADISPA470D
  • Antigen symbol:
  • HSP47
  • HSP47
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • M16.10A1
  • M16.10A1
  • Reactivity:
  • Hamster, Mouse, Pig, Rabbit, Monkey, Sheep, Chicken, Human, Bovine, Guinea pig, Dog, Rat
  • Hamster, Mouse, Pig, Rabbit, Monkey, Sheep, Chicken, Human, Bovine, Guinea pig, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Native rat HSP47
  • Native rat HSP47
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein G affinity chromatography
  • Purified by Protein G affinity chromatography
  • Storage buffer:
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Storage temperature:
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together