Ecolite(+)™ liquid scintillation cocktail, MP Biomedicals
Catalog # ICNA0188247501
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:3,79 L
- Description:Ecolite(+)™ liquid scintillation cocktail
- Environmentally Preferable:
Specifications
About this item
EcoLite(+)™ is a unique formulation of media possessing exceptional water and salt holding characteristics. Ready-to-use scintillation cocktail for a wide variety of aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is essential.
- Environmentally safe
- Non toxic and non flammable
- Ready-to-use
- Versatile
- Excellent stability for RIA and receptor assays
- Forms no gel or phasing
- Ideal for all types of samples
Suitable for difficult-to-count RIA samples, basic samples, phosphate buffer samples, and biological fluids.